Learn More
Medchemexpress LLC Griseofulvin | 126-07-8 | 98.9% | 352.77 | 10 G
SDP
Supplier:  Medchemexpress LLC HY1758310G
Encompass_Preferred
Griseofulvin is an orally active antifungal antibiotic with antitumor activity. It induces apoptosis and G2/M cell cycle arrest in cancer cells. It also exhibits cardiovascular modulatory activity, reducing angina pectoris and relieving hand artery spasm associated with onychomycosis and peripheral vascular diseases like shoulder-hand syndrome.
  • Orally active antifungal antibiotic
  • Antitumor activity
  • Induces apoptosis and G2/M cell cycle arrest in cancer cells
  • Cardiovascular modulatory activity
  • Reduces angina pectoris and relieves hand artery spasm
  • Inhibits growth of various cell lines
  • Increases ventricular contractility and coronary blood flow in animals
  • Reduces tumor volume in xenograft mice
  • Microbial metabolite
  • Microtubule/Tubulin inhibitor
  • Wee1, Caspase, and Bcl-2 Family inhibitor
Catalog No. 50-003-86753