Learn More
Sino Biological Peptide Substrates: GRKtide
Supplier:  Sino Biological G4658
Encompass_Preferred
The synthetic peptide sequence (CRRREEEEESAAA) is routinely evaluated as a substrate for GRK family kinases, including GRK2, GRK3, GRK4 and GRK7.
Catalog No. 50-256-9066