Learn More
Medchemexpress LLC GRL-1720 | 2835511-03-8 | C14H11ClN2O2 | 50 MG

Supplier: Medchemexpress LLC HY15353350MG
GRL-1720 is a potent SARS-CoV-2 Mpro inhibitor with an EC50 value of 15 μM. It exhibits anti-SARS-CoV2 activity and has a molecular weight of 274.70.
- Potent SARS-CoV-2 Mpro inhibitor
- Exhibits anti-SARS-CoV2 activity
- EC50 value of 15 μM (SARS-CoV-2 Mpro)
- Molecular weight of 274.70
- Appearance: off-white to light yellow solid
- Purity: 98.87%
- Protected VeroE6 cells cultured with SARS-CoV-2WK-521
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.