Learn More
Medchemexpress LLC GRO-alpha/CXCL1 Rat 10ug

Supplier: Medchemexpress LLC HYP718910UG
Recombinant rat GRO-alpha (CXCL1) is a 72-amino-acid chemokine produced in E. coli and supplied as a lyophilized powder for research applications. It mediates neutrophil recruitment and angiogenesis and is provided in small research-scale vials, formulated in phosphate buffer with physiological salt, and characterized by high purity for reliable in vitro use.
- Recombinant mature rat CXCL1, 72 amino acids long.
- Expressed in E. coli and supplied lyophilized for stability.
- Formulated in phosphate buffer with physiological salt concentration.
- Purity greater than 95% as determined by reducing SDS-PAGE.
- Predicted mass ~7.9 kDa; apparent ~10 kDa by SDS-PAGE.
- Suitable for biochemical assays, receptor binding, and in vitro functional studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.