Learn More
Enzo Life Sciences GRO-beta (human), (recombinant) (100µg)
SDP
Supplier:  Enzo Life Sciences ENZPRT2370100
Encompass_Preferred
Alternative name: MGSAbeta, CXCL2, MIP-2alpha, GRO2. Purity: ≥95% (SDS-PAGE). Formulation: Lyophilized from a sterile (0.2 micron) filtered solution containing 0.1% Trifluoroacetic Acid (TFA). Source: Produced in E. coli. Contains 73 amino acids. Long Term Storage: -80°C.
Catalog No. 50-201-4039