Learn More
Enzo Life Sciences GroEL (E. coli) monoclonal antibody (9A1/2) (200µg)
SDP
Supplier:  Enzo Life Sciences ENZABS7140200
Encompass_Preferred
Clone: 9A1/2. Isotype: IgG1κ. Immunogen: E. coli GroEL. Source: Purified from hybridoma tissue culture supernatant. Species reactivity: E. coli. Applications: IP, WB. Purity: Protein G affinity purified. Formulation: Liquid. In PBS containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-201-3252