Learn More
Enzo Life Sciences GroEL (E. coli), (recombinant) (25µg)
SDP
Supplier:  Enzo Life Sciences ALX201141C025
Encompass_Preferred
Alternative name: Heat shock protein 60, 60kDa chaperonin, Protein Cpn60 GroEL protein, HSP60. Concentration: 14.1mg/ml. Purity: ≥99%. Formulation: Liquid. In 50mM TRIS/HCl, pH 8.0, containing 10mM KCl and 10mM MgCl2. Source: Produced in E. coli. Long Term Storage: -80°C. Use/Stability: Full activity after short incubation at room temperature. Handling: For maximum product recovery after thawing, centrifuge the vial before opening the cap. Avoid freeze/thaw cycles.
Catalog No. 50-200-7935