Learn More
Grainger Grooving GDM 3020N150RCM PR1535 10 pcs.
Supplier:  Grainger 170NF0
Encompass_Preferred
The Grainger GDM 3020N150RCM PR1535 Grooving Insert is a 10-pack of carbide inserts for grooving and parting. The inserts have a 0.118" width of cut and are made of micro-grain carbide for wear resistance and strength. They feature a 15 degree entering angle and 35 degree front clearance angle. The inserts have an ISO designation of CNMG and are designed for use on steel, stainless steel, cast iron, and non-ferrous materials. The inserts fit holders with an ISO SCLCR/L 1212 designation and have a hole size of 5mm.
Catalog No. 50-276-4448