Learn More
Grainger Grooving KCG 2062R A65 Unc 10 pcs.
Supplier:  Grainger 170NM1
Encompass_Preferred
The 170NM1 is a 1/2 HP, 115/230V, 1725 RPM, 56C Frame, Open Drip Proof, Single Phase, TEFC motor manufactured by Baldor Electric Company. It has a 1.15 service factor, Class F insulation, and a C-Face mounting style. The motor is designed for general purpose applications and features cast iron construction with a thermally protected frame. At 115V it draws 8.4A and at 230V it draws 4.2A. The motor dimensions are 9-1/4" long x 5-7/16" wide x 6-7/16" high and it weighs 26 pounds.
Catalog No. 50-276-4380