Learn More
Cusabio Technology LLC Fish growth hormone,GH ELISA Kit
Supplier:  Cusabio Technology LLC CSBE12121FH
Encompass_Preferred
This Fish growth hormone (GH) ELISA Kit was designed for the quantitative measurement of Fish growth hormone (GH) protein in serum, plasma, tissue homogenates. It is a Competitive ELISA kit, its detection range is 312.5 pg/mL-5000 pg/mL and the sensitivity is 312.5 pg/mL. One kit contains 96 wells.
Catalog No. NC2952939