Learn More
Medchemexpress LLC GRP75-IN-1 dihydrochloride | 2922680-16-6 | 99.75% | 539.60 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY162774A100MG
Encompass_Preferred
GRP75-IN-1 dihydrochloride is an anti-endometrial cancer (EC) agent that induces apoptosis. It reduces Ca2+ levels in mitochondria by targeting GRP75 and disrupting its interaction with IP3R.
  • Induces apoptosis in endometrial cancer cells
  • Reduces mitochondrial Ca2+ levels
  • Targets GRP75 and disrupts its interaction with IP3R
Catalog No. 50-003-92746