Learn More
TARGETMOL CHEMICALS INC Grp94 Inhibitor-1 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T726125MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. Grp94 Inhibitor-1 is a potent and selective inhibitor of Grp94 with an IC50 of 2 nM. Purity 97.02%
Catalog No. 50-288-0140