Learn More
Apexbio Technology LLC GS-441524 1191237-69-0 25mg
SDP
Supplier:  Apexbio Technology LLC B846125
Encompass_Preferred
GS-441524 (CAS 1191237-69-0) is a nucleoside analog targeting viral RNA polymerase It functions by interfering with viral RNA polymerase activity thereby inhibiting viral RNA synthesis and replication GS-441524 exerts its biological activity primarily through inhibition of viral RNA synthesis In cell-based studies GS-441524 demonstrates inhibitory activity with an IC50 value of 0 78 M against feline infectious peritonitis virus Based on these pharmacological properties GS-441524 holds research potential in the treatment of feline infectious peritonitis (FIP)
Catalog No. 50-000-89407