Learn More
Medchemexpress LLC GS-443902 trisodium | 1355050-21-3 | 99.9% | 597.15 | 1 MG
Supplier: Medchemexpress LLC 1MGHY126303C
GS-443902 trisodium is a potent inhibitor of viral RNA-dependent RNA-polymerases (RdRp). It has IC50 values of 1.1 μM for RSV RdRp and 5 μM for HCV RdRp. This compound is the active triphosphate metabolite of Remdesivir, intended for research use only.
- Potent viral RNA-dependent RNA-polymerases (RdRp) inhibitor
- Active triphosphate metabolite of remdesivir
- Inhibits RSV RdRp-catalyzed RNA synthesis
- Inhibits HCV RdRp-catalyzed RNA synthesis
- Selectively inhibits EBOV replication
- Stable under recommended storage conditions
- Store at -80°C, protect from light, and under nitrogen
- Freshly prepared solutions are recommended due to instability