Learn More
Medchemexpress LLC HY-126303C 5mg Medchemexpress, GS-443902 (trisodium) CAS:1355050-21-3 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY126303C5MG
Encompass_Preferred
Medchemexpress, HY-126303C 5mg GS-443902 (trisodium) CAS:1355050-21-3 GS-443902 trisodium (GS-441524 triphosphate trisodium) is a potent viral RNA-dependent RNA-polymerases (RdRp) inhibitor with IC50s of 1.1 µM, 5 µM for RSV RdRp and HCV RdRp, respectively. GS-443902 trisodium is the active triphosphate metabolite of Remdesivir (GS-5734)[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8899