Learn More
Medchemexpress LLC HY-10081 5mg Medchemexpress, GS-6201 CAS:752222-83-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY100815MG
Encompass_Preferred
Medchemexpress, HY-10081 5mg GS-6201 CAS:752222-83-6 GS-6201 (CVT-6883) is a selective adenosine A2B receptor antagonist. GS-6201 displays high affinity and selectivity for the human adenosine A2B receptors (Ki=22 nM). GS-6201 reduces caspase-1 activity in the heart, and attenuates cardiac remodeling after acute myocardial infarction (AMI) in the mouse. GS-62013 attenuates the airway reactivity induced by NECA, AMP, or allergen in sensitized mice[3]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-7783