Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GS-704277 | 1911579-04-8 | MFCD32701941, MFCD22380600 | 99.6% | 442.32 | C15H19N6O8P | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1363035MG

Encompass_Preferred

GS-704277 is the alanine metabolite of the antiviral prodrug remdesivir, provided as a high-purity research reference standard for metabolic, antiviral, and analytical studies. It serves as a metabolic intermediate and analytical standard, and is used to study remdesivir metabolism and downstream antiviral activity.

  • High purity reference standard (99.63%).
  • Molecular formula C15H19N6O8P; molecular weight 442.32.
  • Soluble in DMSO at 100 mg/mL; may require ultrasonication.
  • Suitable for metabolic, antiviral, and analytical research applications.
  • Storage: 4°C under nitrogen; in solvent store -80°C (6 months) or -20°C (1 month).
  • Available in small research quantities, including 5 mg.

Catalog No. 50-002-22555


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.