Learn More
Sino Biological Peptide Substrates: GS Peptide
Supplier:  Sino Biological G6058
Encompass_Preferred
The GS peptide sequence (PLSRTLSVSS) is derived from an N-terminus of glycogen synthase and is suitable for use as the substrate for DCAMKL1 and CAMKII family.
Catalog No. 50-256-9060