Learn More
Medchemexpress LLC GSK-1292263 | 1032823-75-8 | 99.7% | 25 MG

Supplier: Medchemexpress LLC HY1206625MG
GSK1292263 is an orally active GPR119 agonist used in research on glucagon-like peptide-1 secretion and glucose homeostasis relevant to type 2 diabetes mellitus. It is suitable for preclinical and in vitro studies where modulation of GPR119 is required.
- Orally available GPR119 agonist.
- Reported pEC50 6.9 (human) and 6.7 (rat).
- Molecular formula C23H28N4O4S; molecular weight 456.56 g/mol.
- High supplier-reported purity (~99.7%).
- Intended for research use in metabolic disease and glucose homeostasis studies.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.