Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC GSK 1562590 hydrochloride | 1003878-07-6 | 99.0% | 617.95 g/mol | C30H31Cl3N4O4 | 5 MG
SDP

Supplier:  Medchemexpress LLC HY1084465MG

Encompass_Preferred

GSK 1562590 hydrochloride is the hydrochloride salt of a high-affinity, selective urotensin-II (UT) receptor antagonist supplied as a research-grade solid for in vitro and in vivo pharmacology studies.

  • High-affinity, selective urotensin-II receptor antagonist (pKi 9.14-9.66).
  • Hydrochloride salt, white to off-white solid.
  • Purity 99.0% and molecular weight 617.95 g/mol.
  • Soluble in DMSO; formulations for in vivo are soluble to ≥ 2.5 mg/mL.
  • Store protected from light under nitrogen; solid at -20°C, in solution -80°C up to 6 months.
  • Intended for research use only, not for human therapeutic use.

Catalog No. 50-002-63811


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.