Learn More
Medchemexpress LLC Gsk-2256098 hydrochloride | 1416771-10-2 | 99.9% | 451.35 g/mol | C20H24Cl2N6O2 | 1 ML

Supplier: Medchemexpress LLC HY100498A10MM 1ML
A selective focal adhesion kinase (FAK) inhibitor provided as a 10 mM stock solution in DMSO for in vitro research use. Supplied as a sealed 1 mL aliquot with high chemical purity and defined storage recommendations.
- High purity (99.9%).
- Ready-to-use 10 mM stock solution in DMSO.
- Small-volume 1 mL aliquot suitable for screening and assay development.
- Molecular weight 451.35 g/mol; molecular formula C20H24Cl2N6O2.
- Contains CAS 1416771-10-2 for unambiguous identification.
- Store at -80°C up to 6 months or -20°C up to 1 month (sealed, away from moisture).
- For research use only; not for clinical or human use.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.