Learn More
TARGETMOL CHEMICALS INC GSK 2830371 25MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T652125MG
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg and bulk. Please contact Fisher for quotes. GSK 2830371 is an orally active, allosteric Wip1 phosphatase inhibitor with IC50 of 6 nM. Purity 99.3%
Catalog No. 50-287-6888