Learn More
Selleck Chemical LLC GSK 2837808A 5mg
SDP
Supplier:  Selleck Chemical LLC S85905MG
Encompass_Preferred
GSK2837808A is a selective LDHA (lactate dehydrogenase A) inhibitor with IC50 of 2.6 nM and 43 nM for hLDHA and hLDHB, respectively. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-217-3375