Learn More
Selleck Chemical LLC AR-A014418 10mg 487021-52-3 GSK-3 Inhibitor VIII
SDP
Supplier:  Selleck Chemical LLC S743510MG
Encompass_Preferred
AR-A014418 (GSK-3? Inhibitor VIII) is an ATP-competitive, and selective GSK3? inhibitor with IC50 and Ki of 104 nM and 38 nM in cell-free assays, without significant inhibition on 26 other kinases tested. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC0717845