Learn More
Medchemexpress LLC Gsk-3β inhibitor 11 | 536731-65-4 | 98.02% | 393.42 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY14813250MG
Encompass_Preferred
GSK-3β inhibitor 11 (compound 21) is a glycogen synthase kinase-3β (GSK-3β) inhibitor with an IC50 of 10.02 μM.
It can be used in neurodegenerative disease research.
  • Glycogen synthase kinase-3β (GSK-3β) inhibitor
  • IC50 of 10.02 μM
  • Suitable for neurodegenerative disease research
Catalog No. 50-003-63774