Learn More
Medchemexpress LLC GSK-3β inhibitor 13 | 2227316-74-5 | 99.74% | 414.46 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1490545MG
Encompass_Preferred
GSK-3β inhibitor 13 is an orally active and potent GSK-3β inhibitor with blood-brain permeability. It significantly decreases the phosphorylation of tau, which is associated with the formation of neurofibrillary tangles in Alzheimer's disease.
  • Orally active and potent GSK-3β inhibitor
  • Exhibits blood-brain permeability
  • Inhibits GSK-3β with an IC50 of 0.73 nM
  • Inhibits GSK-3α with an IC50 of 0.35 nM
  • Decreases phosphorylation of tau (IC50=58 nM)
Catalog No. 50-003-60927