Learn More
Medchemexpress LLC GSK-3β inhibitor 2 | 1702428-31-6 | ≥95.0% | 318.35 g/mol | C14H14N4O3S | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1307955MG
Encompass_Preferred
GSK-3β inhibitor 2 is a potent, selective, and orally active inhibitor of glycogen synthase kinase-3β (GSK-3β) with an IC50 of 1.1 nM. It is reported to cross the blood-brain barrier and is used in preclinical research to probe GSK-3β-mediated signaling pathways. The compound is supplied as a powder; molecular formula C14H14N4O3S and molecular weight 318.35 g/mol.
  • Potent and selective GSK-3β inhibition (IC50 1.1 nM).
  • Demonstrated blood-brain barrier penetration.
  • Suitable for in vitro and in vivo preclinical studies.
  • Provided as a powder for solution preparation.
  • Molecular formula C14H14N4O3S; molecular weight 318.35 g/mol.
Catalog No. 50-002-62136