Learn More
Medchemexpress LLC Gsk-3β inhibitor 22 | 1005199-84-7 | 99.38% | 423.43 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1649075MG
Encompass_Preferred
GSK-3β inhibitor 22 (compound 20o) is a GSK-3β inhibitor with an IC50 of 3.1 nM. GSK-3β inhibitor 22 has the potential of the study of Alzheimer's disease.
  • GSK-3β inhibitor with an IC50 of 3.1 nM.
  • Potential for the study of Alzheimer's disease.
Catalog No. 50-003-91594