Learn More
TARGETMOL CHEMICALS INC GSK-3 INHIBITOR 4 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T773415MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg and bulk. Please contact Fisher for quotes. GSK-3 inhibitor 4 is an orally active and brain-permeable triple inhibitor of GSK-3 CDK2 and CDK5 with inhibitory effects on GSK-3(beta) GSK-3(alpha) CDK2 and CDK5 with IC50 values of 0.56 nM 0.45 nM 0.47 uM 0.68 uM respectively.GSK-3 inhibitor 4 can effectively reduce Tau protein levels. GSK-3 inhibitor 4 can effectively reduce the level of Tau protein.GSK-3 inhibitor 4 can be used in the study of Alzheimer's disease. purity: 98%
Catalog No. 50-257-9151