Learn More
Medchemexpress LLC HY-10580 5mg Medchemexpress, GSK 3 Inhibitor IX CAS:667463-62-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY105805MG
Encompass_Preferred
Medchemexpress, HY-10580 5mg GSK 3 Inhibitor IX CAS:667463-62-9 GSK 3 Inhibitor IX (6-Bromoindirubin-3'-oxime; BIO) is a potent, selective, reversible and ATP-competitive inhibitor of GSK-3α/β and CDK1-cyclinB complex with IC50s of 5 nM/320 nM/80 nM for (GSK-3α/β)/CDK1/CDK5, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8295