Learn More
Medchemexpress LLC GSK 3008348 hydrochloride | 1629249-40-6 | 99.9% | 524.11 | 50 MG
SDP
Supplier:  Medchemexpress LLC HY19767A50MG
Encompass_Preferred
GSK 3008348 hydrochloride is a small molecule antagonist of integrin αvβ6. It targets the αvβ6 integrin, inhibits TGF-β activation, and can alleviate the fibrotic process. It also prevents the binding of the foot-and-mouth disease virus (FMDV) to the αvβ6 integrin receptor, inhibiting virus entry. GSK 3008348 hydrochloride can be used for research on pulmonary fibrosis and various types of cancer.
  • Small molecule antagonist of integrin αvβ6.
  • Inhibits TGF-β activation.
  • Alleviates the fibrotic process.
  • Prevents binding of foot-and-mouth disease virus (FMDV) to the αvβ6 integrin receptor.
  • Inhibits viral entry.
  • Used for research on pulmonary fibrosis and various types of cancer.
  • Antagonizes integrin αvβ6 with high affinity.
  • Suppresses FMDV RNA replication and virus production.
  • Decreases TGF-β1-activated procollagen I production.
  • Inhibits TGF-β-induced phosphorylation of ERK1/2 and STAT3.
Catalog No. 50-004-01853