Learn More
CPC Scientific H-Lys-Glu-Ala-Pro-Pro-Ala-Pro-Pro-Gln-Ser(PO3H2)-Pro-NH2 (trifluoroacetate salt) 1MG
SDP
Supplier:  CPC Scientific CASP026B
Encompass
ONE-LETTER SEQUENCE: KEAPPAPPQS(PO3H2)-P-NH2MOLECULAR FORMULA: C50H81N14O18P1MOLECULAR WEIGHT:1197.3STORAGE CONDITIONS: -20 5CRESEARCH AREA: ApoptosisREFERENCES: Plotkin, B., et al. 2003. J. Pharmacol. Exp. Ther,. 305, 974.
Catalog No. 50-210-8968