Learn More
Cayman Chemical GSK 417651A 25mg
SDP
Supplier:  Cayman Chemical 1526825 MG
Encompass_Preferred
A TRPC3 and TRPC6 inhibitor (IC50 = ~0.4 µM for both); inhibits migration of isolated human fibroblastic valve interstitial cells into the wounded area in a scratch test assay at 2 µM
Catalog No. 50-297-1269