Learn More
Selleck Chemical LLC GSK 5959
SDP
Supplier:  Selleck Chemical LLC S591625MG
Encompass_Preferred
GSK5959 is a potent and selective BRPF1 bromodomain inhibitor with an IC50 of 80 nM and exhibits 100-fold selectivity for BRPF1 over a panel of 35 other bromodomains including BRPF2/3 and BET family bromodomains
Catalog No. 50-000-03897