Learn More
Apexbio Technology LLC GSK 650394, 5mg. Cas: 890842-28-1 MFCD: MFCD12828779. SGK1 inhibitor
SDP
Supplier:  Apexbio Technology LLC B10515
Encompass_Preferred
GSK 650394 is a small molecule inhibitor of serum and glucocorticoid-regulated kinase 1 (SGK1) with IC50 value of 13nM [1].Sgk1 gene is a target gene of androgen. Knockdown of SGK1 expression attenuates androgen-mediated growth of the prostate cancer cell line. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5202