Learn More
Medchemexpress LLC HY-15192 5mg Medchemexpress, GSK 650394 CAS:890842-28-1 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY151925MG
Encompass_Preferred
Medchemexpress, HY-15192 5mg GSK 650394 CAS:890842-28-1 GSK 650394 is a novel SGK inhibitor with IC50 of 62 nM and 103 nM for SGK1 and SGK2 in the SPA assay respectively. GSK 650394 also inhibits influenza virus replication. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-9093