Learn More
Medchemexpress LLC GSK 690 Hydrochlorid 25mg | 2436760-79-9 | 405.92 | C24H24ClN3O | 25 MG
SDP
Supplier:  Medchemexpress LLC HY117226A25MG
Encompass_Preferred
GSK 690 hydrochloride is a reversible inhibitor of lysine-specific demethylase 1 (LSD1) used as a research reagent in biochemical and cellular studies. It exhibits low nanomolar binding and biochemical potency and is supplied as a small-scale package for laboratory use.
  • Reversible LSD1 inhibitor with Kd 9 nM and biochemical IC50 37 nM.
  • High chemical purity (98.6%) for in vitro applications.
  • Well characterized molecular properties (MW 405.92 g/mol, formula C24H24ClN3O).
  • Intended for biochemical and cellular research use.
Catalog No. 50-002-26774