Learn More
TARGETMOL CHEMICALS INC GSK-872 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T407410MG
Encompass_Preferred
Also available in 1 mg 2 mg 5 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GSK-872 (GSK2399872A) is an effective and specific RIP3 kinase inhibitor. It binds RIP3 kinase domain with high affinity (IC50 1.8 nM) and inhibits kinase activity (IC50 1.3 nM). purity: 99%
Catalog No. 50-257-6816