Learn More
Selleck Chemical LLC GSK 872
SDP
Supplier:  Selleck Chemical LLC S846510MM/1ML
Encompass_Preferred
GSK 872 is a potent and selective RIP3 kinase inhibitor which binds RIP3 kinase domain with high affinity (IC50 1 8 nM) and inhibits kinase activity with IC50 of 1 3 nM It has minimal cross-reactivity
Catalog No. 50-000-02207