Learn More
Medchemexpress LLC GSK-872 hydrochloride | 2703752-81-0 | MFCD30481302 | 99.2% | 419.95 g/mol | C19H18ClN3O2S2 | 1 ML
SDP
Supplier:  Medchemexpress LLC HY101872A10MM 1ML
Encompass_Preferred
GSK-872 hydrochloride is a potent, selective inhibitor of receptor-interacting protein kinase 3 (RIPK3). It binds the RIP3 kinase domain and inhibits kinase activity with IC50 values of 1.8 nM (binding) and 1.3 nM (kinase inhibition). Supplied at high purity as a 10 mM solution in DMSO, the compound is intended for biochemical and cell-based research into necroptosis and RIPK3 signaling.
  • Potent RIPK3 inhibition (IC50 1.3-1.8 nM).
  • High purity suitable for research (>99%).
  • Supplied as a 10 mM solution in DMSO, 1 mL, ready to use.
  • Suitable for biochemical and cell-based assays studying necroptosis.
  • COA and SDS documentation available for quality and safety verification.
Catalog No. 50-002-47938