Learn More
Medchemexpress LLC HY-10299 5mg Medchemexpress, GSK-923295 CAS:1088965-37-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY102995MG
Encompass_Preferred
Medchemexpress, HY-10299 5mg GSK-923295 CAS:1088965-37-0 GSK-923295 is a special, allosteric inhibitor of centromere-associated protein-E (CENP-E) kinesin motor ATPase activity, with Ki of 3.2±0.2 nM and 1.6± 0.1 nM for human and canine, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-8004