Learn More
Selleck Chemical LLC GSK J4 HCl (GSKJ4 HCl) 10mM/1mL 1797983-09-5
SDP
Supplier:  Selleck Chemical LLC S707010MM/1ML
Encompass_Preferred
GSK J4 HCl is a cell permeable prodrug of GSK J1, which is the first selective inhibitor of the H3K27 histone demethylase JMJD3 and UTX with IC50 of 60 nM in a cell-free assay and inactive against a panel of demethylases of the JMJ family. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1001463