Learn More
Cayman Chemical GSK-J4hydrochlorIde 50mg
SDP
Supplier:  Cayman Chemical 1207350 MG
Encompass_Preferred
An ethyl ester prodrug of the JMJD3 selective histone demethylase inhibitor GSK-J1; reduces LPS-induced proinflammatory cytokine production, including that of TNFα (IC50 = 9 µM) in human primary macrophages
Catalog No. 50-295-4177