Learn More
Selleck Chemical LLC GSK-LSD1 2HCl
SDP
Supplier:  Selleck Chemical LLC S7574100MG
Encompass_Preferred
GSK-LSD1 2HCl is an irreversible and selective LSD1 inhibitor with IC50 of 16 nM 1000 fold selective over other closely related FAD utilizing enzymes (i e LSD2 MAO-A MAO-B)
Catalog No. 50-000-04574