Learn More
Selleck Chemical LLC GSK-LSD1 2HCl 5mg 2102933-95-7
SDP
Supplier:  Selleck Chemical LLC S75745MG
Encompass_Preferred
GSK-LSD1 2HCl is an irreversible, and selective LSD1 inhibitor with IC50 of 16 nM, > 1000 fold selective over other closely related FAD utilizing enzymes (i.e. LSD2, MAO-A, MAO-B). *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4964