Learn More
Selleck Chemical LLC GSK0660
SDP
Supplier:  Selleck Chemical LLC S581725MG
Encompass_Preferred
GSK0660 is a potent PPAR / antagonist with a pIC50 of 6 8 (binding assay IC50 155 nM antagonist assay IC50 300 nM) and is nearly inactive on PPAR and PPAR with IC50s above approximately 10 M
Catalog No. 50-000-03741