Learn More
Selleck Chemical LLC GSK1016790A
SDP
Supplier:  Selleck Chemical LLC S810725MG
Encompass_Preferred
GSK1016790A (GSK101) is a novel potent activator of TRPV4 (transient receptor potential vanilloid 4) with EC50 of 34nM in choroid plexus epithelial cells
Catalog No. 50-000-00544