Learn More
Medchemexpress LLC GSK1016790A | 942206-85-1 | 99.9% | 655.61 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY196085MG
Encompass_Preferred
GSK1016790A is a potent and selective transient receptor potential vanilloid 4 (TRPV4) channel agonist. It can elicit Ca2+ influx and elevate intracellular Ca2+ in HEK cells.
  • Elicits Ca2+ influx in mouse and human TRPV4-expressing human embryonic kidney (HEK) cells (EC50 values of 18 and 2.1 nM, respectively).
  • Evokes a dose-dependent activation of TRPV4 whole-cell currents at concentrations above 1 nM.
  • Treatment elicits a rapid elevation of intracellular Ca2+ in a subset of neurons.
  • Produces a dose-dependent inhibitory effect on whole gut transit time in mice.
  • Significantly inhibits electrical field stimulation (EFS)-induced twitch contractions in isolated mouse colon strips in a concentration-dependent manner.
Catalog No. 50-004-01270