Learn More
Apexbio Technology LLC GSK1070916, 100mg Cas# 942918-07-2
SDP
Supplier:  Apexbio Technology LLC A4127100
Encompass_Preferred
reversible and ATP-competitive Aurora B/C inhibitor 942918-07-2. MFCD16039223 GSK1070916 is a potent and selective inhibitor of Aurora B/C Kinase with Ki values of 0.38nM and 1.5nM, respectively [1]. There are other sizes available. Please inqury us for quote.
Catalog No. 50-199-8503