Learn More
Selleck Chemical LLC GSK1070916 S2740-5mg
SDP
Supplier:  Selleck Chemical LLC S27405MG
Encompass_Preferred
GSK1070916 is a reversible and ATP-competitive inhibitor of Aurora B/C with IC50 of 3 5 nM/6 5 nM It displays 100-fold selectivity against the closely related Aurora A-TPX2 complex Phase 1
Catalog No. 50-217-3098